Learn More
Corning™ Flex Discharge Tubes
Corning™ Flex Discharge Tubes for quick and precise dispensing of acids, bases and solvents
$48.75 - $75.47
Specifications
| Product Type | Flexible Discharge Tube |
|---|---|
| Description | Coiled |
| Material | FEP, Polypropylene |
| For Use With (Application) | Quick and precise dispensing of acids, bases and solvents |
| Includes | Protective Sleeve |
Description
- Coiled
- Includes protective sleeve
Specifications
| Flexible Discharge Tube | |
| FEP, Polypropylene | |
| Protective Sleeve |
| Coiled | |
| Quick and precise dispensing of acids, bases and solvents |