missing translation for 'onlineSavingsMsg'
Learn More

Flecainide acetate, 98%

Catalog No. p-7042722
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63527MC 100 mg
AAJ6352703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63527MC Supplier Thermo Scientific Chemicals Supplier No. J63527MC
Only null left

Sodium channel blocker | CAS: 54143-56-5 | C19H24F6N2O5 | 474.4 g/mol

Flecainide acetate is used as a sodium channel blocker and as an anti-arrhythmic. In general, it is used to regulate heartbeats of the upper heart and lower heart chambers. It can also be used to slow down an overactive heart. It is also useful in the treatment of ventricular tachycardias and ventricular arrhythmias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute acetic acid, dimethyl sulfoxide, ethanol, methanol and water.

Notes
Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 54143-56-5
Molecular Formula C19H24F6N2O5
Molecular Weight (g/mol) 474.4
MDL Number MFCD00214290
InChI Key RKXNZRPQSOPPRN-UHFFFAOYSA-N
Synonym flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
PubChem CID 41022
ChEBI CHEBI:5091
IUPAC Name acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
SMILES CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F

Specifications

Melting Point 145°C to 147°C
Color White
Quantity 100 mg
Merck Index 14,4098
Solubility Information Soluble in dilute acetic acid,dimethyl sulfoxide,ethanol,methanol and water.
Formula Weight 474.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Flecainide acetate
Hazard Category H302-H315-H319-H335-H361
Hazard Statement GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 258-997-5
RTECSNumber CV5792550
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only