missing translation for 'onlineSavingsMsg'
Learn More

Flavone 98.0+%, TCI America™

Catalog No. p-7129441
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
F00151G 1 g
F00155G 5 g
2 options

Catalog No. F00151G

Supplier: TCI America F00151G

Only null left

Chemical Identifiers

CAS 525-82-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00006825
InChI Key VHBFFQKBGNRLFZ-UHFFFAOYSA-N
Synonym flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril
PubChem CID 10680
ChEBI CHEBI:42491
IUPAC Name 2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2

Specifications

Melting Point 95°C
Color White
Boiling Point 185°C
Quantity 1 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Flavone
RTECSNumber DJ3100630
TSCA Yes