missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavanone 98.0+%, TCI America™
$41.02 - $117.43
Chemical Identifiers
| CAS | 487-26-3 |
|---|---|
| Molecular Formula | C15H12O2 |
| Molecular Weight (g/mol) | 224.26 |
| MDL Number | MFCD00006841 |
| InChI Key | ZONYXWQDUYMKFB-UHFFFAOYNA-N |
| Synonym | flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one |
| PubChem CID | 10251 |
| ChEBI | CHEBI:5070 |
| IUPAC Name | 2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one |
| SMILES | O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1 |
Chemical Identifiers
| 487-26-3 | |
| 224.26 | |
| ZONYXWQDUYMKFB-UHFFFAOYNA-N | |
| 10251 | |
| 2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one |
| C15H12O2 | |
| MFCD00006841 | |
| flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one | |
| CHEBI:5070 | |
| O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1 |
Specifications
| 487-26-3 | |
| White-Yellow | |
| MFCD00006841 | |
| flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one | |
| O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1 | |
| 224.26 | |
| CHEBI:5070 | |
| ≥98.0% (GC) | |
| Flavanone |
| 76°C | |
| C15H12O2 | |
| 5 g | |
| ZONYXWQDUYMKFB-UHFFFAOYNA-N | |
| 2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one | |
| 10251 | |
| 224.26 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : LK6906500
TSCA : Yes