missing translation for 'onlineSavingsMsg'
Learn More

Flavanone 98.0+%, TCI America™

Catalog No. F02555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02555G 5 g
F025525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02555G Supplier TCI America Supplier No. F02555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 487-26-3
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00006841
InChI Key ZONYXWQDUYMKFB-UHFFFAOYNA-N
Synonym flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one
PubChem CID 10251
ChEBI CHEBI:5070
IUPAC Name 2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one
SMILES O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Flavanone
RTECSNumber LK6906500
TSCA Yes