missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Flat Tooth Picks - Pack of 1000

Catalog No. S02901
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02901 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02901 Supplier Eisco™ Supplier No. TPFLATPK1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compostable 3 1/2 in. wooden tooth pick - 1000 per bag.

  • Made from high-quality wood
  • Perfect for crafting and educational projects
  • The smooth, flat finish ensures safety and ease of use
  • Ideal for skewering small items, detailed work, and precise applications
  • Eco-friendly and compostable

Specifications

Grade K to 12
Format Kit
Product Type Flat Tooth Pick
Subject Science