missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ FlaskScrubber Analytical Laboratory Kit

Catalog No. 09311206
Change view
Click to view available options
Dimensions (L x W x H):
19.2 x 17.8 x 8.7 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
09311206 19.2 x 17.8 x 8.7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09311206 Supplier Labconco™ Supplier No. 4577400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Outfit a FlaskScrubber to produce contaminate-free labware for use in sensitive analytical techniques.

  • Use to produce contaminate-free labware for use in sensitive analytical techniques
  • Includes 15 x 45 mm Vial Insert (4574500), 12 x 32 mm Vial Insert (4574400), 24-Place Pipette Insert (4575400), Graduated Cylinder Insert (4574600), and Upper Standard Rack (4668500)

Specifications

Material Stainless Steel
Product Type Starter Kit
Color Silver
Rack Type Spindle, Standard
Style Analytical Starter Kit
For Use With (Application) Washing Glassware
For Use With (Equipment) FlaskScrubber Glassware Washer
Includes 15 x 45 mm Vial Insert (4574500), 12 x 32 mm Vial Insert (4574400), 24-Place Pipette Insert (4575400), Graduated Cylinder Insert (4574600), and Upper Standard Rack (4668500)
Dimensions (L x W x H) 19.2 x 17.8 x 8.7 in.
Weight (English) Shipping 47 lb.
Weight (Metric) Shipping 21.3 kg
Show More Show Less