Learn More
Chemglass Inc Volumetric Flask, 500 +/- 0.20mL Capacity, #19 Stopper Size, Approx 275mm Height with Stopper
Supplier: Chemglass Inc CG1616500
Volumetric Flask, 500 +/- 0.20mL Capacity, #19 Stopper Size, Approx 275mm Height with Stopper. Low actinic red stained glass provides protection to light sensitive solutions. Allowing less than 1% transmission below 400nm and 5% from 400-600nm. Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique Flat Bottom Interior permits the use of a standard stir bar. Large permanent white letters make the capacity clearly visible. Flasks are calibrated "To Contain" at Class A tolerances, per ASTM E288 specifications. Supplied complete with a glass stopper.