Learn More
Chemglass Inc 10mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Glass Stopper
Supplier:  Chemglass Inc CG160002
Encompass
10mL Volumetric Flask, Class A, #9 Outer Stopper Neck, Glass Stopper. Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class A unserialized flasks and ASTM E-288. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.
Catalog No. CG160002
Only null left