Learn More
BUCHI Flask Seals for Industrial Rotavapor
Catalog No. 05402571
Change view
Click to view available options
Type:
Evaporating Flask Seal, Complete
Catalog No. Type
05402571 Evaporating Flask Seal, Complete
1 options
Catalog No. 05402571 Supplier BUCHI Supplier No. 041121
Original BUCHI seal for Industrial Rotavapor. Optimize the performance of your distillation with high quality and chemically resistant replacement parts.
  • High chemical resistance
  • OEM seal for proper fit and performance
  • PTFE, Viton and silicone seal
Specifications
Type Evaporating Flask Seal, Complete
For Use With (Equipment) Flask seals