missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Flask Holder

Catalog No. 15050321
Change view
Click to view available options
:
Flask Holder, 8 x 5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
15050321 Flask Holder, 8 x 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15050321
Only null left

This Flask Holder is constructed from 16 gauge, type 304 stainless steel with a #4 finish.

  • This Flask Holder is constructed from 16 gauge, type 304 stainless steel with a #4 finish.
  • This is autoclavable.
  • Ships with (2) SP-4 pegs and (2) HP-1 hole plugs to mount on pegboard.

Specifications

Product Type Flask Holder
Depth (English) 5 in.
Width (English) 8 in.