Learn More
Fisherbrand™ Flask Covers
Catalog No. p-127557
Change view
Click to view available options
Closure Size:
6
7 - 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
10042B 7 - 8
10042A 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10042B
Only null left
Cut down on evaporation.
  • Flint glass
  • Designed by Professor F.E. Tuttle
  • Place in necks of flasks while solutions are being boiled to cause condensation and refluxing
  • Use with any flask that takes rubber stopper sizes listed in ordering table
Specifications
Closure Material Flint Glass
Material Glass
For Use With (Equipment) Use with flask that takes Rubber stopper sizes, used when solutions are being boiled to cause condensation and refluxing
Product Type Cover, Flask
Closure Size 7 - 8