Learn More
BUCHI FlashPure Select Flash Chromatography Cartridges: Silica
Catalog No. 05410084
Change view
Click to view available options
FlashPure Select; Stationary Phase: Silica; Size: 120 g, Quantity: 10 pcs
Catalog No.
05410084 FlashPure Select; Stationary Phase: Silica; Size: 120 g, Quantity: 10 pcs
1 options
Catalog No. 05410084 Supplier BUCHI Supplier No. 140000118
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FlashPure Select cartridges are designed for ultimate and unmatched performance.
  • Spherical particles
  • 13-17 μm silica, 20-35 μm C18
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4 g - 3,000 g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Material Polypropylene
Type Silica
Includes 10 Pc.
Resolution 16 to 24 μm
Size (Metric) Tube Rack 120 g
Column Type Silica
For Use With (Application) Difficult Separations of Polar Compounds
For Use With (Equipment) Any Flash System