missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure Select Flash Chromatography Cartridges: C18

Catalog No. 05410094
Change view
Click to view available options
Size (Metric) Tube Rack:
4 g
12 g
20 g
40 g
80 g
120 g
220 g
330 g
Includes:
1 Pc.
2 Pc.
5 Pc.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Tube Rack Includes
05410094 330 g 1 Pc.
05410092 120 g 2 Pc.
05410088 12 g 5 Pc.
05410089 20 g 2 Pc.
05410093 220 g 1 Pc.
05410090 40 g 2 Pc.
05410087 4 g 5 Pc.
05410091 80 g 2 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 05410094 Supplier BUCHI Supplier No. 140000128

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FlashPure Select cartridges are designed for ultimate and unmatched performance.

  • Spherical particles
  • 13-17 μm silica, 20-35 μm C18
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4 g - 3,000 g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.

Specifications

Material Polypropylene
Type C18
Includes 1 Pc.
Resolution 20 to 35 μm
Size (Metric) Tube Rack 330 g
Column Type C18
For Use With (Application) Non-Polar Compounds, Ideal for Peptides
For Use With (Equipment) Any Flash System