Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Alumina Neutral
Catalog No. 05410073
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
FlashPure EcoFlex; Stationary Phase: Alumina Neutral; Size: 160g, Quantity: 10pcs
Catalog No.
05410073 FlashPure EcoFlex; Stationary Phase: Alumina Neutral; Size: 160g, Quantity: 10pcs
1 missing translation for 'options'
Catalog No. 05410073 missing translation for 'mfr' BUCHI missing translation for 'supplierNo' 140000099
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Material Polypropylene
Type Neutral Alumina
Includes 10 Pc.
Resolution 50 to 75 μm
Size (Metric) Tube Rack 160 g
Column Type Alumina Neutral
For Use With (Application) Polar Compounds
For Use With (Equipment) Any Flash System