Learn More
BUCHI FlashPure Chromatography Solid Loader
Catalog No. 05410130
Change view
Click to view available options
FlashPure Empty Solid Loader; Size: 120 g; Frits: Included; Quantiry: 10 pcs
Catalog No.
05410130 FlashPure Empty Solid Loader; Size: 120 g; Frits: Included; Quantiry: 10 pcs
1 options
Catalog No. 05410130 Supplier BUCHI Supplier No. 140000177
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Type Empty Cartridge
Includes Frits
Size (Metric) Tube Rack 120 g