missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: C18

Catalog No. 05410027
Change view
Click to view available options
Includes:
1 Pc.
2 Pc.
5 Pc.
Bed Weight (Metric):
4 g
12 g
20 g
40 g
80 g
120 g
220 g
330 g
800 g
1500 g
3000 g
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Bed Weight (Metric)
05410027 2 Pc. 120 g
05410023 5 Pc. 12 g
05410031 1 Pc. 1500 g
05410024 2 Pc. 20 g
05410028 1 Pc. 220 g
05410032 1 Pc. 3000 g
05410029 1 Pc. 330 g
05410025 2 Pc. 40 g
05410022 5 Pc. 4 g
05410030 1 Pc. 800 g
05410026 2 Pc. 80 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 05410027 Supplier BUCHI Supplier No. 140000053

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.

  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters

Specifications

Material Polypropylene
Type C18
Includes 2 Pc.
Resolution 40 to 60 μm
Bed Weight (Metric) 120 g
Column Type C18
For Use With (Application) Non-Polar Compounds, Ideal for Peptides
For Use With (Equipment) Any Flash System