Learn More
Corepoint™ Scientific Flammable Material Storage Manual Defrost Touchscreen Freezers
Elevate your lab safety with Corepoint™ Flammable Storage Freezers.
Supplier:  Corepoint™ Scientific FF171WWW/0MHCTS
 View more versions of this product
Catalog No. 361058664
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Flammable storage freezers meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. Utilizes non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
  • Intuitive Touchscreen Controllers
  • Microproessor temperature control
  • Temperature alarms and datalogging
  • Probe access
  • Keyed door locks
  • Natural hydrocarbon refrigerants
  • Our freezers showcase elite features including state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Chemical Identifiers
Freezer
Solid Swing
Stainless Steel
Stainless Steel
6
Upright
White
3-year Parts and Labor Warranty, 5-year Compressor Parts Warranty
Audible and Visual High and Low Temperature Alarms
115 V
R290
-15°C to -25°C
Scratch-resistant Painted Steel
EPA-compliant High-density Urethane Foam
Powder-coated Steel
ETL, NFPA, ENERGYSTAR
No
Fan-forced
Manual
NEMA 5-15
H-drive
Specifications
Freezer
481 L
Solid Swing
Stainless Steel
Stainless Steel
6
Upright
White
3-year Parts and Labor Warranty, 5-year Compressor Parts Warranty
Audible and Visual High and Low Temperature Alarms
NEMA 5-15
H-drive
17 cu. ft
-15°C to -25°C
Scratch-resistant Painted Steel
EPA-compliant High-density Urethane Foam
Powder-coated Steel
ETL, NFPA, ENERGYSTAR
No
Fan-forced
Manual
6 A
115 V
R290
Product Suggestions
Product Suggestions