Learn More
National Safety Apparel™ Flame Resistant Grid Fleece Cap
Catalog No. 17001699
Encompass
Change view
Click to view available options
Color: Navy
Catalog No.
17001699 Color: Navy
1 options
Catalog No. 17001699 Supplier National Safety Apparel™ Supplier No. H01FWCAP
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For added warmth and FR protection, the flame resistant grid fleece cap offers a lightweight breathable option to keep you comfortable on and off the job site. With flat lock seams for maximum comfort, four-way stretch and high performance moisture wicking technology this cap will be your go-to as the weather transitions through the cooler months.
  • Arc Rating: 15 cal/cm²
  • CAT 2
  • Fabric/Material: 9.6 oz. FR POLARTEC™ POWER GRID™ Fleece
  • Four-way stretch for ultimate comfort and flexibility
  • Grid fabric technology allows superior breathability
  • High performance moisture wicking
  • Warmth without the weight
  • Flat lock seams for maximum comfort
  • Made in the USA
Specifications
Size Medium
Material POLARTEC™ POWER GRID™ Fleece
Color Navy
Seam Type Stitched
Arc Flash Rating 15 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462