missing translation for 'onlineSavingsMsg'
Learn More

FL-70™ Concentrate

Catalog No. SF1054
Change view
Click to view available options
Volume (English):
33.87 oz.
1.05 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
SF1054 1.05 gal.
SF1051 33.87 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SF1054 Supplier Thermo Fisher Scientific Supplier No. SF105 4
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powerful alkaline detergent removes dirt, grime, organic residue, coagulated blood, dried gelatin and stopcock grease

  • Contains no polyphosphates or chromates to interfere with analysis
  • Biodegradable
  • Approx. 10pH
  • Use 2% solution for normal jobs and 5% for more difficult tasks

Specifications

Form Liquid
pH 10
Appearance Yellow
Boiling Point >100°C
Composition Tetrasodium Ethylenediamineteraacetate 1.4%, Sodium Oleate 0.5%, Sodium Bicarbonate 0.1%, Sodium Car
For Use With (Application) Removes dirt, grime, organic residue, coagulated Blood, dried Gelatin and stopcock grease
Volume (English) 1.05 gal.