Recombinant Protein;Human FKBP12 / FKBP1A Protein;1MG;Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contains AA Met 1 - Glu 108 (Accession # AAH05147).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.8 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human FKBP12 / FKBP1A Protein;1MG;Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contains AA Met 1 - Glu 108 (Accession # AAH05147).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.8 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).