missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fixture Cutout for Custom Worksurface

Catalog No. 06000390
Catalog No. 06000390
Only null left

This Fixture cutout is a unit price for a custom work surface that requires fixture cut outs.

  • Contact your Fisher representative for details

Specifications

Location Custom
Color Black
For Use With (Application) Laboratory Environmnet