missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fixogum Rubber Cement

Catalog No. MP1FIXO0125
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1FIXO0125 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1FIXO0125 Supplier MP Biomedicals™ Supplier No. 11FIXO0125

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridization for FISH.

Applications: Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridization for FISH.

Specifications

Content And Storage Room Temperature
Format Tube
Product Type Biochemicals
Quantity 125 mL
Form Liquid

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.