missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveGo™ F2 pH/mV Meters

Catalog No. p-7070339
Change view
Click to view available options
Type:
Meter with Field Kit
Meter with Food Kit
Meter with Standard Kit
pH/mV Meter
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01912356 pH/mV Meter
01912357 Meter with Standard Kit
01912358 Meter with Field Kit
01912359 Meter with Food Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01912356 Supplier Mettler Toledo™ Supplier No. 30266946

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack

Portable meter provides reliable and high quality pH/mV measurements with simple click of a button

Different combinations of portable meters, sensors and accessories are available, which are ideally suited for specific applications or samples.

  • Robust, IP67 waterproof ABS housing
  • Large, well-structured 3.1 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • Ergonomic design provide comfort to both big and small hands
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • Withstands wet and demanding environments
  • Includes compact carrying case
  • Three point calibration with four predefined buffer groups
  • ATC and MTC temperature compensation
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • BNC, Cinch sensor inputs
  • Ideal for mobile applications in the laboratory, at-line, or outdoors
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01mV
  • mV Range: -1999 to 1999mV
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: 0° to +100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.5°C
  • Memory Size: 200 measurements, current calibration
  • Operating Hours: >200 hrs.
  • Power Supply: 4 x 1.5V AAA LRO3 or 4 x 1.2V AAA NiMH HR03
  • Dimensions: 18.8 x 7.7 x 3.3cm (97.4 x 3 x 1.3 in.)
  • Weight: 0.26kg (0.5 lb.)
Applications:
  • F2-Meter only: Suitable for applications that require a specific sensor not included in a kit
  • F2 Food Kit: Suitable for solid and semi-solid samples such as meat, fruit, cheese, and more
  • F2 Field Kit: Suitable for wide range of general aqueous samples and for measurements on the go
  • F2 Standard Kit: Suitable for wide range of general aqueous samples including emulsions and suspensions

Specifications

Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
Includes Electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack
For Use With (Application) Applications that require a specific sensor not included in a kit
Measurement Modes pH
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) -1999 to 1999
Resolution (mV) 1mV
Calibration Points 3
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°
Display Type LCD
Backlight Option None
Memory 200 measurements, current calibration
Data Points 200
Log Function Type Automatic, Manual
Inputs BNC, Cinch
Dimensions (L x W x H) 18.8 x 7.7 x 3.3 cm (97.4 x 3 x 1.3 in.)
Weight (Metric) 0.26 kg
Weight (English) 0.5 lb.
Model F2
Temperature Compensation ATC and MTC
Type pH/mV Meter
Show More Show Less