missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveGo™ F2 pH/mV Meters

Catalog No. 01912357
Change view
Click to view available options
Type:
Meter with Field Kit
Meter with Food Kit
Meter with Standard Kit
pH/mV Meter
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01912357 Meter with Standard Kit
01912356 pH/mV Meter
01912358 Meter with Field Kit
01912359 Meter with Food Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01912357 Supplier Mettler Toledo™ Supplier No. 30266889
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

LE438 IP67 sensor, electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack

Portable meter provides reliable and high quality pH/mV measurements with simple click of a button

Different combinations of portable meters, sensors and accessories are available, which are ideally suited for specific applications or samples.

  • Robust, IP67 waterproof ABS housing
  • Large, well-structured 3.1 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • Ergonomic design provide comfort to both big and small hands
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • Withstands wet and demanding environments
  • Includes compact carrying case
  • Three point calibration with four predefined buffer groups
  • ATC and MTC temperature compensation
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • BNC, Cinch sensor inputs
  • Ideal for mobile applications in the laboratory, at-line, or outdoors
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01mV
  • mV Range: -1999 to 1999mV
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: 0° to +100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.5°C
  • Memory Size: 200 measurements, current calibration
  • Operating Hours: >200 hrs.
  • Power Supply: 4 x 1.5V AAA LRO3 or 4 x 1.2V AAA NiMH HR03
  • Dimensions: 18.8 x 7.7 x 3.3cm (97.4 x 3 x 1.3 in.)
  • Weight: 0.26kg (0.5 lb.)
Applications:
  • F2-Meter only: Suitable for applications that require a specific sensor not included in a kit
  • F2 Food Kit: Suitable for solid and semi-solid samples such as meat, fruit, cheese, and more
  • F2 Field Kit: Suitable for wide range of general aqueous samples and for measurements on the go
  • F2 Standard Kit: Suitable for wide range of general aqueous samples including emulsions and suspensions

Specifications

Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
Includes LE438 IP67 sensor, electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack
For Use With (Application) For wide range of general aqueous samples including emulsions and suspensions
Measurement Modes pH
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) -1999 to 1999
Resolution (mV) 1mV
Calibration Points 3
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°
Display Type LCD
Backlight Option None
Memory 200 measurements, current calibration
Data Points 200
Log Function Type Automatic, Manual
Inputs BNC, Cinch
Dimensions (L x W x H) 18.8 x 7.7 x 3.3 cm (97.4 x 3 x 1.3 in.)
Weight (Metric) 0.26 kg
Weight (English) 0.5 lb.
Model F2
Temperature Compensation ATC and MTC
Type Meter with Standard Kit
Show More Show Less