missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters

Catalog No. p-7070335
Change view
Click to view available options
Type:
Meter with Bio Kit
Meter with Micro Kit
Meter with Standard Kit
Meter with TRIS Kit
pH Meter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01912350 Meter with Micro Kit
01912348 Meter with Standard Kit
01912351 Meter with TRIS Kit
01912349 Meter with Bio Kit
01912347 pH Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01912350 Supplier Mettler Toledo™ Supplier No. 30266940

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

LE422 sensor, installation QuickGuide, pH buffer starter pack

Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • RS232 or USB ports to export data directly either to a printer or a PC for further processing
  • ATC and MTC temperature compensation
  • Five point (max.) linear only calibration with four predefined buffer groups
  • BNC, Cinch, NTC 30kiloOhm sensor inputs
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • pH Range: -2.00 to 16.00
  • pH Resolution: 0.01/0.1
  • pH Accuracy: ±:0.01
  • mV Range: 2000 to 2000mv
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: -5° to +105°C (23° to 221°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.3°C
  • Memory Size: 200 measurements, current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • FP20 Meter only: Suited for applications that require a specific sensor not included in a kit
  • FP20 Standard Kit: Suited for wide range of general aqueous samples
  • FP20 Bio Kit: Suited for fast results in aqueous samples such as acids, bases
  • FP20 Micro Kit: Suited for small samples in vials and test tubes
  • FP20 TRIS Kit: Suited for demanding, dirty samples such as emulsions, oily samples, waste water, TRIS

Specifications

Description FiveEasy Plus FP20-Micro
Accuracy (mV) ±1
Accuracy (Temperature) 0.3°C
Accuracy (pH) ±0.01
Data Management Details RS232, USB
Includes FP20 pH meter, LE422 pH sensor, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide
For Use With (Application) For small samples in vials and test tubes
Calibration Linear/Segmented
Range (pH) -2 to 16
Resolution (pH) 0.01, 0.1
Range (mV) 2000 to 2000
Resolution (mV) 1mV
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1°C
Display Type LCD
Memory 200 measurements, current calibration
Inputs BNC, Cinch, NTC 30kiloOhm sensors
Dimensions (L x W x H) 22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
Weight (Metric) 0.63 kg
Weight (English) 1.4 lb.
Model FP20
Probe Type LE422 pH sensor
Temperature Compensation ATC and MTC
Type Meter with Micro Kit
Voltage 120/240 V
Frequency 50/60 Hz
Show More Show Less