missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ F30 Conductivity Meters

Catalog No. p-7070338
Change view
Click to view available options
Type:
pH Meter
pH Meter with Standard Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01912352 pH Meter
01912353 pH Meter with Standard Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01912352 Supplier Mettler Toledo™ Supplier No. 30266942

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Installation QuickGuide, 1413μS/cm and 12.88mS/cm standard sachets

Compact, benchtop meter provides high quality conductivity measurements with simple click of a button

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • One point calibration with three predefined standards
  • Mini-DIN sensor input
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • Conductivity Range: 0.01μS/cm to 200mS/cm
  • Conductivity Resolution: Automatic range
  • Conductivity Accuracy: ±0.5% of the measured value
  • TDS Range: 0.01mg/L to 200g/L
  • TDS Resolution: Automatic range
  • TDS Accuracy: ±0.5% of the measured value
  • Temperature Range: 0° to 100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.5°C
  • Temperature Compensation: Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
  • Memory Size: current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • F30 Meter only: Suited for applications that require a specific sensor not included in a kit
  • F30 Standard Kit: Suited for aqueous samples with mid to high conductivity

Specifications

Description FiveEasy F30-Basic
Accuracy (Conductivity) ±0.5% of the measured value
Includes F30 pH meter, electrode holder, 1 each buffers 1413mS/cm and 12.88mS/cm standard sachets, test certificate
For Use With (Application) Applications that require a specific sensor not included in a kit
Measurement Modes Conductivity
Calibration One point calibration with three predefined standards
Range (Conductivity) 0.01 μS/cm to 200 mS/cm
Resolution (Conductivity) Automatic range
Temperature Compensation Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
Range (Salinity) 0 to 42 psu
Range (TDS) 0 mg/L to 300 mg/L
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 32°F to 212°F
Resolution (Temperature) 0.1°C
Accuracy (Temperature) 0.5°C
Display Type LCD
Memory Current calibration
Data Points 200
Inputs Mini-DIN
Dimensions (L x W x H) 22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
Weight (Metric) 0.63 kg
Weight (English) 1.4 lb.
Model F30
Type pH Meter
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less