missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ F20 pH/mV Meters

Catalog No. p-7070336
Change view
Click to view available options
Type:
Meter with Standard Kit
pH Meter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01912345 pH Meter
01912346 Meter with Standard Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01912345 Supplier Mettler Toledo™ Supplier No. 30266658

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

F20 pH meter, electrode holder, pH buffer starter pack, test certificate, pH measurement guide. (Electrode not included.)

Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • ATC and MTC temperature compensation
  • Three point (max.) linear calibration with four predefined buffer groups
  • BNC, Cinch, NTC 30kiloOhm sensor inputs
  • Features analog output, Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Accuracy: ±:0.01
  • mV Range: 2000 to 2000mv
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: 0° to +100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.5°C
  • Memory Size: current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • F20 Meter only: Suited for applications that require a specific sensor not included in a kit

Specifications

Description FiveEasy F20-Basic
Includes F20 pH meter, electrode holder, 1 buffer sachets each for pH 4.01, 7, 9.21, and 10.01, test certificate, pH measurement guide
Accuracy (Temperature) 0.5°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
For Use With (Application) Applications that require a specific sensor not included in a kit
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) 2000 to 2000
Resolution (mV) 1mV
Temperature Range (Metric) 0°C to 100°C
Temperature Range (English) 32°F to 212°F
Resolution (Temperature) 0.1°C
Display Type LCD
Memory Current calibration
Inputs BNC, Cinch, NTC 30kiloOhm sensors
Dimensions (L x W x H) 22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
Weight (Metric) 0.63 kg
Weight (English) 1.4 lb.
Model F20
Temperature Compensation ATC and MTC
Type pH Meter
Voltage 120/240 V
Frequency 50/60 Hz
Show More Show Less