Learn More
GSC Go Science Crazy Five Metal Heat Conductometer
Catalog No. S13644
Change view
Click to view available options
Dimensions (L x W):
12 x 8 in.
Catalog No. Dimensions (L x W)
S13644 12 x 8 in.
1 options
Intended for ages 13 and up.
Catalog No. S13644 Supplier GSC Go Science Crazy Supplier No. 431201
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Intended for ages 13 and up.
Includes
Aluminum, steel, brass, copper, & nickel rods; Wax for visualization; Instructions
Demonstrates the different conductive properties of various metals. The GSC International Five Metal Heat Conductometer contains one spoke each of aluminum, steel, brass, copper, and nickel extending from a central hub. This tool is perfect for classroom demonstrations of heat conduction principles.
  • Used to demonstrate the different conduction properties of various metals
  • Consists of five different metal spokes that extend from a center hub which is connected to a wooden handle
  • When the hub is heated, students watch as wax falls from each spoke in the order of highest conductivity
  • Ideal for classroom or other educational settings
  • Wax and instructions included
  • Metals include aluminum, steel, brass, copper, and nickel
Specifications
Dimensions (L x W) 12 x 8 in.
For Use With (Application) Testing conduction properties of various metals