missing translation for 'onlineSavingsMsg'
Learn More

FisherTab™ TT-50 Kjeldahl Tablets, Fisher Chemical

Catalog No. k3161000
Change view
Click to view available options
Quantity:
1000 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K3161000 1000 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K3161000 Supplier Thermo Fisher Scientific Supplier No. FLK3161000
Only null left

Composition per tablet: 5g K2SO4 + 0.1g CuSO4 + 0.1g TiO2

Specifications

Chemical Name or Material FisherTab™ TT-50 Kjeldahl Tablets
Color Blue
CAS 13463-67-7, 7778-80-5, 7758-98-7
Physical Form Powder/Solid
Quantity 1000 Tablets
Assay Percent Range 94%
Packaging Poly Bottle
MDL Number MFCD00010981