missing translation for 'onlineSavingsMsg'
Learn More

FisherTab™ ST-AUTO Kjeldahl Tablets, Fisher Chemical

Catalog No. k3131000
Change view
Click to view available options
Quantity:
1000 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K3131000 1000 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K3131000 Supplier Thermo Fisher Scientific Supplier No. FLK3131000
Only null left

Composition per tablet: 1.5g K2SO4 + 0.015g Se

Specifications

Chemical Name or Material FisherTab™ ST-AUTO Kjeldahl Tablets
Color Green
CAS 7782-49-2, 7778-80-5
Physical Form Solid
Quantity 1000 Tablets
Assay Percent Range 99%
Packaging Poly Bottle
MDL Number MFCD03456802