Learn More
Fisherbrand™ Histoplast Paraffin Wax
Three blends to accommodate all specimen types
Supplier: Fisherbrand™ 22900701
Description
When purity matters, only the finest will do.
- Formulated using raw materials of only the highest purity
- Is not sticky or brittle while sectioning
- Provides consistent tissue ribbons every time
Histoplast PE
For general use and all histology applications
Histoplast IM
For use on specimens requiring more thorough infiltration (large and dense specimens)
Histoplast LP
A lower melting-point formula for improved ribboning with less compression
Chemical Identifiers
| 1kg |
Specifications
| 54° to 57°C | |
| Paraffin, Histoplast IM | |
| 1kg | |
| For large and dense specimens |