missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fisherfinest™ Chemically Resistant Markers

Catalog No. 22026700
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22026700 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22026700 Supplier Fisherbrand™ Supplier No. ES001F
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Permanent marker for use on cassettes and the frosted area of microscope slides

 

  • Ultra-fine tip
  • Smearproof & Waterproof
  • Not ethanol resistant
  • Odorless
  • Resistant to most histology chemicals
  • Does not readily dry out
  • Available in black

 

Specifications

Product Type Chemically Resistant Permanent Marker
Tip Style Ultrafine
Color Black
For Use With (Application) For use on cassettes and the frosted area of microscope slides
Quantity 12 Pk.