missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Water- or Air-Driven Turbine Magnetic Safety Stirrer

Catalog No. 14511210
Change view
Click to view available options
Capacity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14511210 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14511210

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

6 ft. (1.8m) length of vinyl tubing for inlet and outlet and fitting for connecting stirrer to pressure line.

Eliminates heat, shock and explosion hazards.

Nonelectric magnetic stirrer with turbine rotor.

  • Propelled by air or water pressure between 15 and 40psi (103 and 276kPa)
  • Corrosion-proof inert plastic throughout
  • Easy to hook up
  • Can be clamped to benchtop or stand with 0.5 in. dia. (13mm) rod

Order Info

Dimensions: 5 dia. x 2.625 in. H (13 x 6.7cm)

Specifications

Speed 800 rpm
Top Plate Material Polypropylene
Capacity 500 mL
Diameter (English) Outside 5 in.
Diameter (Metric) Outside 13 cm
Height (English) 2.625 in.
Height (Metric) 6.7 cm
Material ABS Plastic
Type Magnetic Stirrer