Learn More
Fisherbrand™ Viscometer Holders
Description
For use with both calibrated and uncalibrated Cannon™-Fenske Routine, Cannon-Fenske opaque, Cannon-Manning semimicro and Cannon-Manning vacuum viscometers.
- Available in either molded plastic or neoprene rubber to fit holes with a 2 in. diameter (5.1cm)
- Plastic holders feature spring clip to hold viscometer securely
Specifications
Specifications
| Product Type | Viscometer Holder |
| For Use With (Equipment) | Calibrated and Uncalibrated Cannon-Fenske Routine, Cannon-Fenske Opaque and Cannon Manning Semimicro Viscometers |
| Material | Molded Plastic |