Learn More
Fisherbrand™ Viscometer Holders
Catalog No. 136177
Change view
Click to view available options
Material:
Molded Plastic
Neoprene, Rubber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
136177 Neoprene, Rubber
136176 Molded Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 136177 Supplier Fisherbrand™ Supplier No. 9726M53
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with both calibrated and uncalibrated Cannon™-Fenske Routine, Cannon-Fenske opaque, Cannon-Manning semimicro and Cannon-Manning vacuum viscometers.
  • Available in either molded plastic or neoprene rubber to fit holes with a 2 in. diameter (5.1cm)
  • Plastic holders feature spring clip to hold viscometer securely
Specifications
Product Type Viscometer Holder
For Use With (Equipment) Calibrated and Uncalibrated Cannon-Fenske Routine, Cannon-Fenske Opaque and Cannon Manning Semimicro Viscometers
Material Neoprene, Rubber