missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ UriSystem™ Caps, Blue

Catalog No. 14377258
Change view
Click to view available options
Color:
Blue
Orange
Turquoise
Violet
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14377258 Yellow
14377254 Blue
14377255 Orange
14377256 Violet
14377257 Turquoise
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14377258

Choose from five colors

  • Tight-fitting plastic caps for standard UriSystem tubes (14-377-252 and 14-377-253)
  • Prevent spilling and contamination

Specifications

Product Type UriSystem Cap
Material Plastic
Color Yellow
For Use With (Application) Prevents spilling and contamination
For Use With (Equipment) For standard UriSystem tubes