missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Urine Sedimentation System: Tube

Catalog No. 14375561
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14375561 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14375561 Supplier Fisherbrand™ Supplier No. 14375561

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Fisherbrand™ Urine Sedimentation System is designed to produce the same results as the most popular style urine sedimentation system using the same test procedure.

  • 15mL capacity urine tube graduated to 12mL

Specifications

Description Graduated to 12mL
Type Urine Tube
For Use With (Application) Designed to produce the same results as the most popular style Urine sedimentation system using the same test procedure
Material Polystyrene
Packaging Type Cs. of 500
Quantity 15 mL