Learn More
Fisherbrand™ Tool Holder for Ultrasonic Units
Catalog No. 2940666
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
2940666 Stainless Steel
1 options
Catalog No. 2940666 Supplier Fisherbrand™ Supplier No. 2070760000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use this Fisherbrand™ Tool Holder with Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. Easily support and handle tools within the ultrasonic cleaner.
  • Multiple prongs for to support multiple tools
  • Dual handles easily secures holder in the ultrasonic cleaner
  • Stainless steel
  • Light and easy to maneuver
Specifications
Product Type Tool Holder
For Use With (Equipment) Elmasonic EP20H E-Plus Ultrasonic Cleaner (107-1653), or the FB11203 Advanced Ultrasonic Cleaner
Height (English) 3.6 in.
Length (English) Bottom L: 4.5 in.; Top L: 8.2 in.
Material Stainless Steel
Width (English) Bottom W: 4.5 in.; Top W: 3 in.