Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Supplier: Fisherbrand™ 1003287
Description
- Easy-to-open and close cover
- Muffles noise level
- Designed to be used as drip basin when flipped over
- Chemical-resistant plastic
- Bring your ultrasonic cleaner up to temperature more quickly
- Contain and prevent spills and leakage
Specifications
| Lid | |
| Ultrasonic Cleaner, 28 L | |
| Medical Laboratories and in Industrial Production Processes | |
| Plastic |