missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Acid Resistant Plastic Tubs for Ultrasonic Cleaners

Provide additional tank protection when using harsh chemical cleaning solutions in the ultrasonic cleaner.

Supplier:  Fisherbrand™ 2070900000

 View more versions of this product

Catalog No. 2940675


Only null left

Description

Description

 

  • Designed to provide additional protection against harsh chemical cleaning solutions
  • Comes with a custom-fit cover to prevent leaks and spills
  • External D x W x H:  305 x 365 x 170 mm
  • Internal D x W x H: 190 x 250 x 165 mm

 

Specifications

Chemical Identifiers

Acid Resistant Tub
Plastic cover
Plastic

Specifications

Acid Resistant Tub
Ultrasonic Cleaners: S 100, S 100/H; S 120, S 120/H
Plastic
5.8 L
Plastic cover
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions