missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Acid Resistant Plastic Tubs for Ultrasonic Cleaners

Provide additional tank protection when using harsh chemical cleaning solutions in the ultrasonic cleaner.

Supplier:  Fisherbrand™ 2070890000

 View more versions of this product

Catalog No. 2940674


Only null left

Description

Description

 

  • Designed to provide additional protection against harsh chemical cleaning solutions
  • Comes with a custom-fit cover to prevent leaks and spills
  • External D x W x H: 200 x 300 x 115 mm
  • Internal D x W x H: 95 x 195 x 110 mm

 

Specifications

Chemical Identifiers

Acid Resistant Tub
Plastic cover
Plastic

Specifications

Acid Resistant Tub
Ultrasonic Cleaners: S 30, S 30/H; S 40, S 40/H; S 60, S 60/H
Plastic
1.3 L
Plastic cover
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions