missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ General Purpose Ultrasonic Cleaning Solution

Catalog No. 1533580
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
1533580 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1533580 Supplier Fisherbrand™ Supplier No. CPN 955 006

Cleans a wide range of contaminants.

 

  • Specially suited for cleaning plastics, glass, ceramics, rubber, and all types of metals including aluminum and magnesium
  • High-performance, heavy-duty ultrasonic cleaner formulation
  • Fast and long lasting
  • Odorless, ammonia-free and nonflammable with no acid, caustic or fuming problems
  • Safe to handle, mild on hands
  • Dilute in ratios up to 10 parts water to one part solution
  • In 1 gal. (3.8L) bottle

 

Specifications

Form Liquid
Appearance Blue
Composition Water 70 to 90%, Pentasodium Triphosphate 10 to 20%, Sodium Bicarbonate 5 to 15%, Sodium Lauryl Sulf
For Use With (Application) For cleaning Plastics, Glass, Ceramics, Rubber and all types of Metals
Volume (English) 1 gal.