Learn More
Fisherbrand™ Ultra Flex Clamping Systems
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
$401.47 - $1,119.97
Specifications
For Use With (Equipment) Ultra flex clamping system components
View More Specs
Products 6
Catalog Number Mfr. No. Size Includes Price Quantity & Availability  
Catalog Number Mfr. No. Size Includes Price Quantity & Availability  
02217007
View Documents Promotion Details
Fisherbrand™
80862753
12 in. (Arm) Base Plate
Each for $1,119.97
Only null left
02217011
View Documents Promotion Details
Fisherbrand™
80862756
18 in. (Arm) Bench Clamp
Each for $911.63
Only null left
02217009
View Documents Promotion Details
Fisherbrand™
80862754
13 in. (Arm) Lab Frame Connector
Each for $709.33
Only null left
02217010
View Documents Promotion Details
Fisherbrand™
80862755
18 in. (Arm) Base Plate
Each for $1,037.20
Only null left
02217012
View Documents Promotion Details
Fisherbrand™
80862757
18 in. (Arm) Lab Frame Connector
Each for $849.53
Only null left
02217016
View Documents Promotion Details
Fisherbrand™
80862758
12 in. (Arm) Flex Arm Only
Each for $401.47
Only null left
Description
Description
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
  • Ultra Flex comes in three different systems: base plate, lab-frame connector, and bench clamp
  • Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305mm (12 in.) or 457mm (18 in.) length, and utility wrench
  • 2-prong and 3-prong clamp heads are also supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers
Specifications
Specifications
Ultra flex clamping system components