Learn More
Fisherbrand™ Ubbelohde Calibrated Viscometer Tubes::Viscometer Holder
Catalog No. 136145
Change view
Click to view available options
For Use With (Equipment):
13-614 Series Viscometer Tube
Catalog No. For Use With (Equipment)
136145 13-614 Series Viscometer Tube
1 options
Catalog No. 136145 Supplier Fisherbrand™ Supplier No. 97226M82
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
With handle. Suspends one 13-614 Series Viscometer Tube in bath with 2 in. (5.1cm) hole in lid.
Specifications
For Use With (Equipment) 13-614 Series Viscometer Tube
Description Viscometer Holder with Handle