missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.

$137.04 - $182.30

Specifications

Bottle Fill Glycol
Certifications/Compliance ISO 17025, A2LA, NIST
For Use With (Application) Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
Height (English) 0.75 in.
Height (Metric) 19 mm
View More Specs

Products 2
Catalog Number Bottle Size Price Quantity & Availability  
Catalog Number Bottle Size Price Quantity & Availability  
15077720
View Documents Promotion Details
5 mL
Each for $182.30
Only null left
 
0666411
View Documents Promotion Details
10 mL
Each for $137.04
Only null left
Edge
 
Description

Description

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.

  • Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
  • Glass bottle filled with glycol mimics vaccine temperature and fits in trays
  • Probe is sealed in glycol solution which insulates sensor when refrigerator door is opened
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
  • Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • High-impact, chemical-resistant ABS plastic case
  • Temperature range: -50 to +70°C/-58 to +158°F
  • Resolution: 1° accuracy: ±1C
  • Battery:AA Alkaline (1)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It provides traceability to standards provided by NIST.

Specifications

Specifications

Glycol
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
19 mm
70 mm
ABS Plastic
4 oz.
2.75 in.
70°C
-50°C
-50°C to +70°C
Refrigerators/Freezers
±1°C
ISO 17025, A2LA, NIST
0.75 in.
10 in.
25.4 cm
1°C
Fridge/Freezer Digital Thermometer
158°F
-58°F
-58°F to +158°F
Yes
AA Alkaline (1)
Digital LCD
Videos
SDS
Documents

Documents