missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ 10-Hour Stopwatches

Catalog No. 146495
Change view
Click to view available options
Time Range:
10 hr.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Time Range
146495 10 hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 146495

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For general-purpose applications

Fisherbrand™ Traceable™ 10-Hour Stopwatches offer single-action timing, time-in/time-out, continuous timing, and cumulative split functions. For general purpose applications.

  • Resolution: 0.01 seconds
  • Handheld with rugged ABS plastic case
  • Net weight: 2.5 oz. (71g)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Product Type Digital Stopwatch
Time Range 10 hr.
Alarms Audible
Accuracy 5e-06
Resolution 0.01 sec.
Display LCD
Material ABS Plastic
Dimensions (L x W x H) 2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9 cm)
Length (English) 2.5 in.
Length (Metric) 6.35 cm
Width (English) 3 in.
Width (Metric) 7.62 cm
Height (English) 0.75 in.
Height (English) Display 0.375 in.
Height (Metric) 1.9 cm
Height (Metric) Display 0.95 cm
Weight (English) 2.5 oz.
Weight (Metric) 0.07 kg
Includes 4 Year Silver Oxide water resistant, O-ring sealed ABS Plastic case and lanyard
For Use With (Application) Enzyme tests, viscosity measurements, animal studies and other timed lab tests
Certifications/Compliance ISO 17025, A2LA, NIST
Show More Show Less