missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Top Grain Leather Palm Gloves

Catalog No. p-2363621
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Catalog No. Size
19048665A Small
19048665B Medium
19048665C Large
19048665D X-Large
4 options

Catalog No. 19048665A

Supplier: Fisherbrand™ 871563/S

Only null left

Top grain leather construction provides durability for an extended useful life.

  • Gunn cut with leather covering palm, thumb, index finger and fingertips
  • Offer more resistance to absorption of oils than other split leathers

Specifications

Color Natural, Blue, Red
Grade Regular
Hand Specific
Material Cowhide Leather
Product Type General Purpose Gloves
Size Small
Style Gunn
Glove Cuff Color Blue with Red Stripes
Glove Cuff Style Safety
Hem Color Red
Liner Material Cotton
Series 87-1563
Length (English) 9.6 in.
Length (Metric) 24.3 cm
Show More Show Less