missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ TFE Starburst Stirring Heads Kit

Catalog No. 1451374
Change view
Click to view available options
Length (Metric):
Assorted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
1451374 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1451374 Supplier Fisherbrand™ Supplier No. 371885000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Set of five bars in assorted sizes.

  • Includes one each: 9.5, 19, 25, 32 and 38 mm dia.
  • With storage container
  • Shape creates a deep vortex
  • Provides quiet, stable operation
  • Anico magnet encapsulated in FDA grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Specifications

Coating PTFE, FDA-grade, USP Class VI
Shape Cross
Type Magnetic Stir Bar Set
Autoclavable Autoclavable
Quantity 30
Certifications/Compliance ISO 9001:2008
Color White
Diameter (English) Assorted
Length (Metric) Assorted
Material Alnico Magnet
Includes Includes one each: 9.5, 19, 25, 32 and 38mm dia., storage container
Show More Show Less