Learn More
Fisherbrand™ Synthetic Blend String Knit Gloves
Ideal for use in warehousing, parts handling, assembly, and food processing applications. Can also be used as glove liners under unprotected work gloves.
Supplier:  Fisherbrand™ 35C110S
 View more versions of this product
Catalog No. 19-059938
Only null left
Explore available promotions
Description
Description
  • Knit pattern allows breathability with less hand fatigue
  • Reversible design allows replacement of one glove at a time, rather than pairs
  • Color-coded hem denotes size
Specifications
Specifications
String Knit
Yes
Uncoated
ANSI A1
No Texture
Cotton
String Knit Gloves
Non-sterile
Yellow
8.9 in.
3.7 in.
Medium Weight
Industrial
Natural
Reusable
Ambidextrous
Powder Free
Small
Continuous Knit
Medium
22.6 cm
9.3 cm
Product Suggestions
Product Suggestions